missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MT-ND1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149795
Description
MT-ND1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 1 (ND1) binds rotenone and rotenone analogs and might be involved in electron transfer to ubiquinone. Mutations in the ND1 gene may be implicated in several disorders, including Leber hereditary optic neuropathy, Alzheimer's disease and Parkinson disease.Specifications
| MT-ND1 | |
| Polyclonal | |
| Unconjugated | |
| mt-Nd1 | |
| mitochondrially encoded NADH dehydrogenase 1; Mtnd1; MT-ND1; NADH dehydrogenase; NADH dehydrogenase subunit 1; NADH1; NADH-ubiquinone oxidoreductase chain 1; Nd1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4535 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P03886 | |
| mt-Nd1 | |
| Recombinant NADH Dehydrogenase 1 (ND1) (Phe186-Phe270). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody |