Learn More
MSA™ Squirt Gas Bump Testers
Catalog No. 189993614
Change view
Click to view available options
Includes:
11L Gas, 1.3% CH4, 15% O2, 60ppm Carbon Monoxide
Catalog No. Includes
189993614 11L Gas, 1.3% CH4, 15% O2, 60ppm Carbon Monoxide
1 options
Catalog No. 189993614 Supplier MSA™ Supplier No. 814497
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.