missing translation for 'onlineSavingsMsg'
Learn More

MSA™ ALTAIR™ Pro Single-Gas Detector

Catalog No. 19165558
Change view
Click to view available options
Components Measured:
Oxygen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
19165558 Oxygen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19165558 Supplier MSA™ Supplier No. 10074137

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sensor and battery replacement options give this instrument an indefinite service life

Features superior dust and water protection, excellent impact resistance, great RFI performance, single-button access to any function, and an extremely effective alarm system.

  • Highly accurate electrochemical sensors measure concentrations of toxic gas or percent oxygen
  • Information displays on large, clear backlit LCD
  • System automatically records the last 50 alarm or test events; active datalogging records peak value every three minutes
  • Triple alarm system consists of two bright flashing LEDs, a piercing audible alarm, and internal vibrating alarm
  • Adjustable alarm setpoints are offered for LOW and HIGH levels on all units; toxic gas versions also have TWA and STEL alarm points
  • Rubberized armor housing resists dust, water and impact
  • Built-in IR communication port connects detector to MSA infrared adapter to read data or change alarm setpoints
  • Built-in gas delivery adapter for gas response tests
  • Functions in a temperature range of -4° to +122°F (-20° to +50°C); relative humidity, 10 to 95%

Order Info

Dimensions (L x W x H): 1 x 2 x 3.4 in. (2.5 x 5.1 x 8.6cm). Weight (including clip): 4 oz. (113g).

Certifications

Rugged housing is IP67 rated for ingress protection. Classified as Intrinsically Safe for use in hazardous locations in the U.S. and Canada (Class I, Groups A, B, C, D; Class II, Group G); ATEX, II 2 G EEx ia II C T4.

Warranty

Two-year manufacturer's warranty on instrument and listed sensors; two years from date of first use or 30 months from date of manufacture, whichever comes first

Specifications

Alarm Types Audible, visual, vibrating
Components Measured Oxygen
Dimensions (L x W x H) 3.4 x 2 x 1 in. (8.63 x 5.08 x 2.54cm)
Height (English) 1 in.
Height (Metric) 2.54 cm
Humidity Range 10 to 95%
IP Rating IP67
Length (English) 3.4 in.
Length (Metric) 8.63 cm
Measurement Range 0 to 25%
Resolution 0.1%
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Warranty 2 Years
Weight (English) 4 oz.
Weight (Metric) 113g
Width (English) 2 in.
Width (Metric) 5.08 cm
Show More Show Less