missing translation for 'onlineSavingsMsg'
Learn More

MS Calibration Solution API, sodium iodide (2 μg/μL) and cesium iodide (50 ng/μL) in 50/50 2-propanol/water, MilliporeSigma™ Supelco™

Catalog No. 111016967
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016967 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016967 Supplier Merck Emd Millipore Supplier No. 1437930ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Solution for LC-MS calibration.

Specifications

CAS 67-63-0
Density 0.910-0.950 g/mL (at 20°C)
Flash Point 22°C
Type Standard
Quantity 30 mL