missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRRF Monoclonal Antibody (OTI1D5)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526771
Description
MRRF Monoclonal Antibody for Western Blot, IHC (P)
Responsible for the release of ribosomes from messenger RNA at the termination of protein biosynthesis. May increase the efficiency of translation by recycling ribosomes from one round of translation to another. Ota T., Nat. Genet. 36:40-45(2004). Humphray S.J., Nature 429:369-374(2004). Zhang Y., Biochim. Biophys. Acta 1443:245-250(1998).Chemical Identifiers
| MRRF | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96E11 | |
| 92399 | |
| Human recombinant protein fragment corresponding to amino acids 56-262 of MRRF produced in E.coli. | |
| Antibody |
| OTI1D5 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| Mrrf | |
| 2400002D02Rik; mitochondrial ribosome recycling factor; MRFF; MRRF; MTRRF; Ribosome-recycling factor, mitochondrial; ribosome-releasing factor, mitochondrial; RRF | |
| Mrrf | |
| Primary | |
| Affinity chromatography |
Specifications
| MRRF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96E11 | |
| Mrrf | |
| Human recombinant protein fragment corresponding to amino acids 56-262 of MRRF produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D5 | |
| Unconjugated | |
| Mrrf | |
| 2400002D02Rik; mitochondrial ribosome recycling factor; MRFF; MRRF; MTRRF; Ribosome-recycling factor, mitochondrial; ribosome-releasing factor, mitochondrial; RRF | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 92399 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |