missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRP6 Monoclonal Antibody (M6II-21)

Rat Monoclonal Antibody

Supplier:  Invitrogen™ MA135686

Catalog No. 01-671-092

Only null left
Explore available promotions

Description

Description

M6II-21 reacts with MRP6, a 190-200 kD transmembrane protein that is related to the multidrug resistance related protein MRP. Mutations in the MRP6 gene are responsible for the connective tissue disorder Pseudoxanthoma elasticum (PXE). M6II-21 was raised against a bacterial fusion protein of human MRP6, containing amino acids 764-964, spanning the putative 12th transmembrane region as well as predicted internal and external regions of the protein. M6II-21 did not cross-react with the human MDR1, MRP1, MRP2, MRP3, MRP4, MRP5 gene products. Unreactive on standard IHC-P.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). The encoded protein, a member of the MRP subfamily, is involved in multi-drug resistance. Mutations in this gene cause pseudoxanthoma elasticum. Alternatively spliced transcript variants that encode different proteins have been described for this gene.
Specifications

Specifications

MRP6
Monoclonal
Conc. Not Determined
tissue culture supernatant with 1% BSA and 0.1% sodium azide
O95255
ABCC6
ABCC6 aa 764-964.
RUO
368
4°C
Liquid
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
M6II-21
Unconjugated
ABCC6
ABC34; Abcc1b; ABCC6; Anthracycline resistance-associated protein; ARA; ATP binding cassette subfamily C member 6; ATP-binding cassette sub-family C member 6; ATP-binding cassette, subfamily C (CFTR/MRP), member 6; ATP-binding cassette, sub-family C (CFTR/MRP), member 6; ATP-binding cassette, sub-family C, member 6; DCC; dyscalc; Dyscalc1; EST349056; GACI2; liver multidrug resistance-associated protein 6; Mlp1; MLP-1; MOATE; MOAT-E; Mrp6; MRP-like protein 1; Multidrug resistance-associated protein 6; multidrug resistance-associated protein-6; multi-specific organic anion transporter E; PXE; PXE1; URG7; URG7 protein; multidrug resistance-associated protein 6
Rat
1 mL
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions