missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRP3 Monoclonal Antibody (1A2F6)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548645
Description
MRP3 Monoclonal Antibody for Western Blot, ELISA
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in the transport of biliary and intestinal excretion of organic anions. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.Chemical Identifiers
| MRP3 | |
| 1 mg/mL | |
| ELISA, Western Blot, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.05% sodium azide | |
| O15438 | |
| 8714 | |
| Purified recombinant fragment of human MRP3 (AA: 849-963) expressed in E. Coli. | |
| Antibody |
| 1A2F6 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| ABCC3 | |
| ABC31; ABCC3; ATP binding cassette subfamily C member 3; ATP-binding cassette sub-family C member 3; ATP-binding cassette, subfamily C (CFTR/MRP), member 3; ATP-binding cassette, sub-family C (CFTR/MRP), member 3; canalicular multispecific organic anion transporter 2; canicular multispecific organic anion transporter; cMOAT2; DKFZp686E22157; EST90757; MLP2; MLP-2; MOAT-D; Mrp3; MRP-like protein 2; multidrug resistance associated protein; multidrug resistance protein 3; multidrug resistance-associated protein 3; multi-specific organic anion transporter D; organic anion transporter | |
| ABCC3 | |
| Primary | |
| Protein G |
Specifications
| MRP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O15438 | |
| ABCC3 | |
| Purified recombinant fragment of human MRP3 (AA: 849-963) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Immunocytochemistry, Western Blot | |
| 1A2F6 | |
| Unconjugated | |
| ABCC3 | |
| ABC31; ABCC3; ATP binding cassette subfamily C member 3; ATP-binding cassette sub-family C member 3; ATP-binding cassette, subfamily C (CFTR/MRP), member 3; ATP-binding cassette, sub-family C (CFTR/MRP), member 3; canalicular multispecific organic anion transporter 2; canicular multispecific organic anion transporter; cMOAT2; DKFZp686E22157; EST90757; MLP2; MLP-2; MOAT-D; Mrp3; MRP-like protein 2; multidrug resistance associated protein; multidrug resistance protein 3; multidrug resistance-associated protein 3; multi-specific organic anion transporter D; organic anion transporter | |
| Mouse | |
| Protein G | |
| RUO | |
| 8714 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |