Learn More
Brady™ MRO Plus™ Absorbent Rolls
Strong, durable and low linting, making them the perfect go-to products for nearly every industrial setting
Supplier:  Brady™ MRO15DP
 View more versions of this product
Catalog No. 19039118
Only null left
Description
Description
  • Highly versatile, gray absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
  • Medium weight - optimal for most general maintenance needs
  • 3-ply Meltblown Polypropylene construction is designed for general industrial applications
  • Non-perforated absorbents offer the greatest absorbency and better tear resistance in high traffic areas
  • Perforations provide cost control and reduce waste by allowing you to use only what you need
Specifications
Chemical Identifiers
Gray
Polypropylene
Specifications
Gray
24 gal.
Universal Sorbent
1 Ea.
15 in. x 150 ft. (38.1 cm x 45.7m )
Perforated every 12 in. and up the center, Dimpled, Medium-linting
Polypropylene
Overspray leaks and drips, workstations, heavy clean-up and wipe-up , drums - liquid storage, outfalls and discharge ponds
Heavyweight
Product Suggestions
Product Suggestions