missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRLC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150188
Description
MRLC2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
MRLC2 is involved in the the activity of nonmuscle myosin II is regulated by phosphorylation of a regulatory light chain. This phosphorylation results in higher MgATPase activity and the assembly of myosin II filaments.Chemical Identifiers
| MRLC2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O14950, Q3THE2 | |
| 103910, 67938 | |
| Recombinant Myosin Light Chain 12B (MYL12B) (Lys5-Asp172). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Pig | |
| MYL12B | |
| 1500001M02Rik; C77744; MLC-2; MLC20; MLC-2A; MLCB; MLC-B; Mrlc2; MRLC3; Mrlcb; Myl12b; MYLC2B; myosin light chain 12B; myosin light chain, regulatory B; myosin regulatory light chain 12B; myosin regulatory light chain 2; Myosin regulatory light chain 20 kDa; myosin regulatory light chain 2-B, smooth muscle isoform; myosin regulatory light chain MRCL2; myosin regulatory light chain MRLC2; myosin regulatory light chain-B; myosin regulatory light polypeptide 9-like; myosin RLC-B; myosin, light chain 12B, regulatory; RLC; RLC-B; SHUJUN-1 | |
| MYL12B | |
| Primary | |
| Protein A |
Specifications
| MRLC2 | |
| Polyclonal | |
| Unconjugated | |
| MYL12B | |
| 1500001M02Rik; C77744; MLC-2; MLC20; MLC-2A; MLCB; MLC-B; Mrlc2; MRLC3; Mrlcb; Myl12b; MYLC2B; myosin light chain 12B; myosin light chain, regulatory B; myosin regulatory light chain 12B; myosin regulatory light chain 2; Myosin regulatory light chain 20 kDa; myosin regulatory light chain 2-B, smooth muscle isoform; myosin regulatory light chain MRCL2; myosin regulatory light chain MRLC2; myosin regulatory light chain-B; myosin regulatory light polypeptide 9-like; myosin RLC-B; myosin, light chain 12B, regulatory; RLC; RLC-B; SHUJUN-1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 103910, 67938 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O14950, Q3THE2 | |
| MYL12B | |
| Recombinant Myosin Light Chain 12B (MYL12B) (Lys5-Asp172). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Pig | |
| Antibody |