missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MR1 Monoclonal Antibody (6A5B2)
GREENER_CHOICE

Catalog No. FERMA548493
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548493 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548493 Supplier Invitrogen™ Supplier No. MA548493
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

MR1 Monoclonal Antibody for ELISA, Flow, ICC/IF, IHC (P), Western Blot

Antigen-presenting molecule specialized in presenting microbial vitamin B metabolites. Involved in the development and expansion of a small population of T-cells expressing an invariant T-cell receptor alpha chain called mucosal-associated invariant T- cells (MAIT). MAIT lymphocytes are preferentially located in the gut lamina propria and therefore may be involved in monitoring commensal flora or serve as a distress signal. Expression and MAIT cell recognition seem to be ligand-dependent.

Specifications

Antigen MR1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6A5B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene MR1
Gene Accession No. Q95460
Gene Alias Class I histocompatibility antigen-like protein; H2ls; histocompatibility-2 complex class 1-like sequence; Hlals; Major histocompatibility complex class I-related gene protein; major histocompatibility complex, class I-like sequence; major histocompatibility complex, class I-related; MCH class I; MHC class I-like antigen MR-1; MHC class I-like sequence; MHC class I-related gene protein; MHC class-I related-gene protein; MR1; Mr1a
Gene Symbols MR1
Host Species Mouse
Immunogen Purified recombinant fragment of human MR1 (AA: extra(23-302)) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3140
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less