missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MPure™ Total RNA Extraction Kit

Catalog No. 117022160
Change view
Click to view available options
Quantity:
48 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
117022160 48 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 117022160 Supplier MP Biomedicals™ Supplier No. 117022160

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MPure™ Total RNA Extraction Kit used with the MPure-12 instrument for extraction of total RNA from whole blood, blood cells, animal tissue, plant tissue, yeast or cultured cells.

  • Nucleic acids extracted can be used in a number of downstream applications including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP, Southern Blot Analysis.

Specifications

Content And Storage MPure-12 Total RNA Extraction Kit should be stored at room temperature (15°C to 25°C). Do not freeze the reagent cartridges. The kits are stable for 15 months under these conditions.
Format Kit
Sample Type Whole Blood, Yeast, Cultured Cells, Animal Tissue, Plant Tissue, Blood Cells
For Use With (Application) RT-PCR, qPCR, sequencing (NGS), microarray, DDRT PCR, TaqMan™ analysis, northern blot analysis, northern blotting
Includes Proteinase K Solution (40 μL), Lysis Buffer 2 (720 μL), Binding Buffer 1 (1300 μL), Magnetic Bead Solution (800 μL), Washing Buffer 1 (1000 μL), Washing Buffer 2 (1000 μL), Washing Buffer 3 (1000 μL), RNase Free Water (1000 μL), Rnase Free Water (1000 μL), DNase
Product Type RNA Extraction Kit
Quantity 48 Tests
Sample Source Animal and Human Tissues

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.