missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPP1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00004354B01P
Description
Sequence: MTLKASEGESGGSMHTALSDLYLEHLLQKRSRPEAVSHPLNTVTEDMYTNGSPAPGSPAQVKGQEVRKVRLIQFEKVTEEPMGITLKLNEKQSCTVARILHGGMIHRQGSLHVGDEILEINGTNVTNHSVDQLQKAMKETKGMISLKVIPNQQSRLPALQMFMRAQFDYDPKKDNLIPCKEAGLKFATGDIIQIINKDDSNWWQGRVEGSSKESAGLIPSPELQEWRVASMAQSAPSEAPSCSPFGKKKKYKDKYLAKHSSIFDQLDVVSYEEVVRLPAFKRKTLVLIGASGVGRSHIKNALLSQNPEKFVYPVPYTTRPPRKSEEDGKEYHFISTEEMTRNISANEFLEFGSYQGNMFGTKFETVHQIHKQNKIAILDIEPQTLKIVRTAELSPFIVFIAPTDQGTQTEALQQLQKDSEAIRSQYAHYFDLSLVNNGVDETLKKLQEAFDQACSSPQWVPVSWVYSpecifications
| MPP1 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_002436.2 | |
| MPP1 | |
| MPP1 (NP_002427.1, 1 a.a. ∼ 466 a.a) full-length human protein. | |
| RUO | |
| 4354 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| membrane protein, palmitoylated 1, 55kDa | |
| AAG12/DXS552/DXS552E/EMP55/MRG1/PEMP | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |