missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MPHOSPH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120351
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5120351 Supplier Invitrogen™ Supplier No. PA5120351
Only null left

Rabbit Polyclonal Antibody

Positive test controls include: U-251MG, OVCAR3, 293T, HT-29, NIH/3T3, Mouse brain, Rat brain. The target is usually found in the following locations: Cell projection, Cytoplasm, Midbody, Nucleus, axon, centrosome, cytoskeleton, growth cone, microtubule organizing center, nucleolus, nucleoplasm, spindle, spindle pole. Immunogen sequence: KHLLQLKEEE EETNRQETEK LKEELSASSA RTQNLKADLQ RKEEDYADLK EKLTDAKKQI KQVQKEVSVM RDEDKLLRIK INELEKKKNQ CSQELDMKQR TIQQLKEQLN NQKVEEAIQQ YERACKDLNV KEKIIEDMRM TLEEQEQTQV EQDQVLEAKL EEVERLATEL EKWKEKCNDL ETKNNQRSNK EHENNTDVLG KLTNLQDELQ ESEQKYNADR KKWLEEKMML ITQAKEAENI RNKEMKKYAE DRERFFKQQN EMEILTAQLT EKDSDLQKWR EERDQLVAAL EIQLKALISS N.

KIF20B is a plus-end-directed motor enzyme that is required for completion of cytokinesis.

Specifications

Antigen MPHOSPH1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KIF20B
Gene Accession No. Q80WE4, Q96Q89
Gene Alias 33cex; B130024C23; C330014J10Rik; Cancer/testis antigen 90; CT90; Kif20b; Kinesin family member 20B; kinesin-like protein KIF20B; Kinesin-related motor interacting with PIN1; KRMP1; magoo; mitotic kinesin-like protein; mitotic kinesin-related protein; M-phase phosphoprotein 1; MPHOSPH1; MPP1; MPP-1; N-6 kinesin
Gene Symbols KIF20B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1200-1500 of human KIF20B (NP_057279.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240641, 309523, 9585
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less