Learn More
MilliporeSigma mPAGE™ Gel Casting Device Kit
Kit for hand casting mini gels with a 0.75 mm, 1.0 mm or 1.5 mm thickness, includes plates and combs
Supplier:  MilliporeSigma MGCK15M
 View more versions of this product
Catalog No. 11-580-598
Only null left
Explore available promotions
Description
Description
  • The mPAGE™ Gel Casting Device Kits offers a leak-free solution for handcasting up to 2 mini protein gels.
  • Gels cast are 8 cm x 10 cm with a gel thickness of 1.0 mm, 0.75 mm, or 1.5 mm, along with 10 or 15-well options for sample loading.
  • The mPAGE™ Gel Caster has been designed to simplify hand casting gels and to eliminate leakage during the casting process.
Specifications
Specifications
Gel Electrophoresis
Gel Caster
1.5 mm
8.3 x 10.1 cm
Gel Caster Base, Gel Caster Frame, Gel Caster Sealing Gaskets, Gel Releasers, Combs 10 wells (0.75 mm, 1.0 mm or 1.5 mm), Combs 15 wells (0.75 mm, 1.0 mm or 1.5 mm), Mini Spacer Plates (0.75 mm, 1.0 mm or 1.5 mm), Mini Short Plates
Product Suggestions
Product Suggestions