missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Yeast RPM Plasmid Isolation Kit

Catalog No. MP112069400
Change view
Click to view available options
No. of Samples:
100 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Samples
MP112069400 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112069400 Supplier MP Biomedicals™ Supplier No. 112069400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Yeast Rapid Pure Miniprep Plasmid Isolation Kit designed for yeast cells. Can isolate plasmid from bacterial transformants for use in any downstream application (PCR, sequencing, restriction enzyme digestion, labeling, etc).

  • The plasmid isolated from yeast is suitable for PCR and bacterial transformation.
  • The same kit can be used to isolate plasmid from bacterial transformants for use in any downstream application (PCR, sequencing, restriction enzyme digestion, labeling, etc).
  • Plasmid DNA mini-preps from yeast rescue positive prey plasmids after a yeast two-hybrid or a yeast one-hybrid screen.

Specifications

Sample Type Yeast Plasmid Prep
For Use With (Application) For PCR and bacterial transformation
Includes Yeast lysing matrix, Alkaline Lysis solution, neutralizing solution, spin Buffer, wash solution concentrate, spin filters, catch tubes, Pre-lysis Buffer
No. of Samples 100 Preps
Sample Volume 1.5 mL
Sample Source Yeast Culture

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.