Learn More
MP Biomedicals™ RPMI 1640 Medium, Powder
General purpose enriched media with extensive applications for a vast range of mammalian cells including human myeloma, mouse hybridoma, human leukocytes, and B and T lymphocytes
Supplier: MP Biomedicals™ 091060122
Specifications
| RPMI 1640 Medium | |
| 1 x 10 L | |
| 4°C |
| No Sodium Bicarbonate | |
| 7.5 to 8.5 |