missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Raffinose Powder
Pentahydrate powder is 98% pure
Supplier: MP Biomedicals™ 114010022
Description
- White powder
Yeast cell culture, Gene expression
Specifications
| 15°C to 30°C | |
| Powder | |
| 454 g |
| D-(+)-Raffinose Pentahydrate | |
| Yeast cell culture, gene expression |