missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Proteinase K

Catalog No. p-3179312
Change view
Click to view available options
Quantity:
1 mg
25 mg
100 mg
250 mg
500 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19350491 1 mg
ICN19350425 25 mg
ICN19350480 100 mg
ICN19350483 250 mg
ICN19350490 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN19350491 Supplier MP Biomedicals™ Supplier No. 0219350491

Suitable for both protein and nucleic acid isolation

Proteinase K, [39450-01-6]
Proteolytic Activity: >30mAnson Units/mg
Unit Definition: One Anson unit is the amount of enzyme which liberates 1 μmol of Folin-positive amino acids per minute at pH 7.5 and 35°C, using hemoglobin as substrate

  • From Tritirachium album
  • Chromatographically purified
  • Exhibits proteolytic activity on proteins, peptides, glycoproteins, amides and esters.
  • Active with nitroanilides of amino acids with protected amino groups, excluding arginine.
  • Useful in the isolation of DNA and RNA, in the analysis of membrane structures and protein structure.

Specifications

For Use With (Application) Protein Structure Analysis
Molecular Weight (g/mol) ∽30
Name Proteinase K
pH Range 7.5 to 12
Purification Method Chromatography
Quantity 1 mg
Source Tritirachium album Limber
Storage Requirements 4°C
Form Lyophilized