missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Plant Culture Salt Mixtures
Plant Culture Salt Mixtures are for the molecular testing of plants and plant-derived samples for improving the productivity, quality, and disease resistance of plants.
Supplier: MP Biomedicals™ 092623020
Description
- Ready to use
- Liquid formulation, prepared according to the original specification, except where noted
- Available in different pack sizes, ready to use and liquid formulations
Specifications
4°C | |
Murashige and Skoog Basal Salt Mixture | |
3.5 to 4.5 | |
Soluble in water |
Powder | |
Solution | |
10 x 1 L |