missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Peptone I from Bovine Tissue
Used in cultivation for indole production and diptheroids
$667.75 - $2339.67
Specifications
| Content And Storage | 15°C to 30°C |
|---|---|
| Form | Powder |
| Product Type | Peptone |
| For Use With (Application) | Cultivation of for indole production and Cultivation of diptheroids |
| Includes | High amount of Tryptophan |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10480601
|
MP Biomedicals™
0210480601 |
1800 g |
Each for $667.75
|
|
|||||
|
ICN10480605
|
MP Biomedicals™
0210480605 |
2270 g |
Each for $2,339.67
|
|
|||||
Description
Water-soluble product, is a mixture of polypeptides and amino acids formed by the partial hydrolysis of protein.
- Format: Cream powder
Cell biology
Specifications
| 15°C to 30°C | |
| Peptone | |
| High amount of Tryptophan |
| Powder | |
| Cultivation of for indole production and Cultivation of diptheroids |