missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Peptone I from Bovine Tissue
Used in cultivation for indole production and diptheroids
Supplier: MP Biomedicals™ 0210480601
Description
Water-soluble product, is a mixture of polypeptides and amino acids formed by the partial hydrolysis of protein.
- Format: Cream powder
Cell biology
Specifications
| 15°C to 30°C | |
| Peptone | |
| High amount of Tryptophan |
| Powder | |
| Cultivation of for indole production and Cultivation of diptheroids | |
| 1800 g |