Learn More
MP Biomedicals™ Pepstatin A
Synthetic inhibitor for pepsin, renin, cathepsin D, and other acid proteases.
Supplier:  MP Biomedicals™ 0219536880
 View more versions of this product
Catalog No. ICN19536880
Only null left
Explore available promotions
Description
Description
Pepstatin A, [26305-03-3]; Isoval-Val-Val-Sta-Ala-Sta
C34H63N5O9, F.W. 685.9; Melting Point: 235-237°C
Specifications
Specifications
Enzyme Assay, Inhibition Assay, Molecular Biology, Protein Purification, Sample Preparation
Pepstatin A
0°C
685.91
100 mg
Powder
Product Suggestions
Product Suggestions