missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lipoprotein-Cholesterol Concentrate
From bovine serum
$513.82 - $513.82
Specifications
| Concentration | 1000 to 2000mg/dL |
|---|---|
| For Use With (Application) | Media Nutrient Supplement, Calibration of Cholesterol Values, Enzymatic or Non-enzymatic Cholesterol Determinations |
| Name | Lipoprotein Cholesterol concentrate |
| pH Range | 7.5 to 8 |
| Source | Bovine Serum |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19147625
|
MP Biomedicals™
0219147625 |
25 mL |
Each for $513.82
|
|
|||||
Description
A mixture of lipoprotein cholesterol, phospholipids and fatty acids. For use as a serum-free or serum-reduced media nutrient supplement, calibration of cholesterol values, and enzymatic or nonenzymatic cholesterol determinations. Offered as a low-salt solution. No preservatives. Free of enzyme inhibitors. Filtered through 0.2μm membrane filter. Cholesterol concentration: 1000-2000mg/dL.
Specifications
| 1000 to 2000mg/dL | |
| Lipoprotein Cholesterol concentrate | |
| Bovine Serum | |
| Solution |
| Media Nutrient Supplement, Calibration of Cholesterol Values, Enzymatic or Non-enzymatic Cholesterol Determinations | |
| 7.5 to 8 | |
| 0° to 5°C |