missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ LB-Agar, Powdered
Offered in a variety of formats for convenience and ease of use
$209.88 - $3224.13
Specifications
| Form | Powder |
|---|---|
| Product Type | Agar |
| For Use With (Application) | For growth of bacteria |
| Catalog Number | Mfr. No. | Quantity | Form | Content And Storage | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Form | Content And Storage | Price | Quantity | |||
|
MP113002232
|
MP Biomedicals™
113002232 |
1000 g | Powder | 15°C to 30°C |
Each for $392.04
|
|
|||
|
MP113002222
|
MP Biomedicals™
113002222 |
454 g | Powder | 15°C to 30°C |
Each for $209.88
|
|
|||
|
MP113002231
|
MP Biomedicals™
113002231 |
1000 g | Powder | 15°C to 30°C |
Each for $611.06
|
|
|||
|
MP113002241
|
MP Biomedicals™
113002241 |
5000 g | Powder | 15°C to 30°C |
Each for $3,224.13
|
|
|||
Specifications
| Powder | |
| Agar | |
| For growth of bacteria |