missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ LB-Agar, Powdered
Offered in a variety of formats for convenience and ease of use
Supplier: MP Biomedicals™ 113002222
Specifications
| Molecular biology Certified™ | |
| Powder | |
| Agar | |
| For growth of bacteria |
| 15°C to 30°C | |
| Molecular biology | |
| Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L | |
| 454 g |