missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ L-Broth Agar, Capsules
For the growth of E.coli and certified for plasmid and other bacterial cultures in liquid medium
$370.20 - $617.14
Specifications
| Content And Storage | 15° to 30°C |
|---|---|
| Product Type | Agar |
| Sterility | Sterile |
| pH | 6.5 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
MP113001221
|
MP Biomedicals™
113001221 |
454 g |
Each for $370.20
|
|
|||||
|
MP113001231
|
MP Biomedicals™
113001231 |
1Â kg |
Each for $617.14
|
|
|||||
Specifications
| 15° to 30°C | |
| Agar | |
| Sterile | |
| 6.5 |