Learn More
MP Biomedicals™ Kanamycin Sulfate
Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis
Supplier:  MP Biomedicals™ 0219453101
 View more versions of this product
Catalog No. ICN19453101
Only null left
Explore available promotions
Description
Description
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Antibiotic effective against most gram positive and gram negative bacteria and most types of yeast.
Specifications
Specifications
15° to 30°C
Crystalline powder
Antibiotic
1 g
Product Suggestions
Product Suggestions