missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Immunidiffusion (RID) Plates

Catalog No. ICN421651
Change view
Click to view available options
Quantity:
12 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN421651 12 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN421651 Supplier MP Biomedicals™ Supplier No. 08421651

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for double and single radial immunodiffusion techniques

  • Polystyrene construction with close-fitting covers to minimize gel dehydration
  • Comes with recessed bottoms to prevent scratching on bench surfaces
  • Hydrophilic in nature for easy gel pouring and better adherence

Specifications

Material Polystyrene
Type Immunodiffusion Plate
For Use With (Application) Double and single radial immunodiffusion techniques
Quantity 12 Pk.
Storage Requirements 15° to 30°C