missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Hemoglobin from Bovine Erythrocytes

Acts as oxygen transporter

Supplier:  MP Biomedicals™ 0210071425

 View more versions of this product

Catalog No. ICN10071425


Only null left
Explore available promotions

Description

Description

Hemoglobin is the major component of red blood cells, and is responsible for their red color. A major oxygen-transporting component of red blood cells Its normal concentration in erythrocytes is 34%. Hemoglobin is the most important respiratory protein of vertebrates by virtue of its ability to transport oxygen from the lungs to body tissues, and to facilitate the return transport of carbon dioxide. Hemoglobin is a tetramer composed of 2 pairs of polypeptide chains called globins, and 4 heme groups. Each polypeptide chain is bound to one heme. Iron is coordinated to 4 pyrrole nitrogens of protoporphyrin IX and to an imidazole nitrogen of a histidine residue from the globin side of the porphyrin.

  • CAS Number 9008-02-0
  • MDL Number MFCD00131282
  • Format: Reddish-brown crystalline powder
  • Moisture content: <10%
  • pH: 7.0 to 9.0 (2% solution)
  • Soluble in water (1 part in 7 of water) and slowly soluble in glycerol

Used to study paradigm for regulation of gene expression and cellular differentiation.

Specifications

Specifications

Cell Biology and Analysis, Cell Culture, Genomics, Molecular Biology
Hemoglobin
25 g
4°C
∽64.5
7 to 9
Bovine
Crystalline
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions