missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Freund's Adjuvant, Complete

Catalog No. ICN642851
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN642851 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN642851 Supplier MP Biomedicals™ Supplier No. 08642851

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enhances the antibody response of animals injected with antigen. Made by a modification of Freund's technique

This product is made by a modification of Freund's technique, in which 25 mg of Mycobacterium is suspended in a mixture of 7.5 mL Arlacel A and 42.5 mL paraffin oil. An adjuvant is a material that when administered enhances the antibody response of animals injected with antigen.

Specifications

Formulation Paraffin oil 85%, Arlacel A 15%, heat-killed Mycobacterium tuberculosis 0.05%
For Use With (Application) Used to enhance the antibody response of animals injected with antigen
Content And Storage 2°C to 8°C
Product Type Freund's Complete Adjuvant
Quantity 50 mL