missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastRNA™ Pro Soil-Direct Kit

Catalog No. MP116070050
Change view
Click to view available options
Quantity:
50 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116070050 50 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116070050 Supplier MP Biomedicals™ Supplier No. 116070050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

RNase Erase™ recommended, FastPrep™ Instrument or alternative high speed disruptor, 100% Ethanol, Microcentrifuge, Chilled 70% Ethanol (prepared with DEPC-treated H2O), Chilled Isopropanol, LRNase-free Microcentrifuge

FastRNA™ Pro Soil-Direct Kit is designed to efficiently isolate total RNA from organic material found in soil samples. Suitable for RT-PCR.

  • Product Overview: The FastRNA™ Pro Soil-Direct Kit is designed to efficiently isolate total RNA from organic material found in soil samples.
  • Application: FastRNA™ Pro Soil-Direct Kit is used to isolate total RNA from organic material found in soil samples. Total RNA isolated is suitable for RT-PCR analysis and other downstream applications.
  • Sample Source: Microorganisms
  • Sample Type: Feces, Soil, Wastewater, Environmental Water, Sludge
  • Sample Volume: Up to 500 mg per prep

Specifications

Sample Type Feces, Soil, Wastewater, Environmental Water, Sludge
For Use With (Application) Nucleic Acid isolation and purification, Protein isolation, molecular biology, gene expression, microbiological analysis
Includes Soil Lysis Solution, (1:1) Phenol: Chloroform, Inhibitor Removal Solution, RNAMATRIX™, Slurry, Binding Solution, Wash Solution Concentrate, DEPC-treated Water, Lysing Matrix E 50
No. of Reactions 50 Preps
Matrix Lysing Matrix E
Product Type FastRNA-24 Kits
Quantity 50 x 2 mL

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.