missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ FastRNA™ Pro Green Kit, 2 mL Tubes
FastRNA™ Pro Green Kit is designed to efficiently isolate total RNA from any type of plant and animal tissue or cultured cells.
Supplier: MP Biomedicals™ 116045050
Requires : FastPrep Instrument or alternative high speed disruptor, microcentrifuge, pipettmen, RNase Erase, Chloroform, 100% ethanol, 75% ethanol, 1.5 or 2.0 mL RNase-free microcentrifuge tubes, Agarose, Gel loading dye and RNA size marker
Description
- Product Overview: The FastRNA™ Pro Green Kit is designed to efficiently isolate total RNA from any type of plant and animal tissue or cultured cells. Using the FastPrep-24™ or FastPrep™ FP120 Instrument, between 50 to 500 mg of tissue can be homogenized by Lysing Matrix D in impact-resistant 2 mL tubes. Total RNA is released into the proprietary, protective RNApro™ Solution. Following extraction with chloroform and precipitation, total RNA is extracted.
- Applications: FastRNA™ Pro Green Kit is used to isolate total RNA from any type of plant and animal tissue or cultured cells. Extracted RNA is suitable for RT-PCR analysis and other downstream applications.
- Sample Type: Animal Tissues, Plants, Cultured Cells
- Sample Volume: Up to 500 mg per prep
Specifications
| Animal Tissue, Plants, Cultured Cells, Insects, Worms | |
| RNApro™ Solution (1 x 55 mL Bottle), DEPC-H2O (1 x 15 mL Bottle), Lysing Matrix D (50 x 2 mL Tubes) | |
| Lysing Matrix D | |
| 50 x 2 mL |
| Nucleic Acid isolation and purification, Protein isolation, molecular biology, gene expression, microbiological analysis | |
| 50 Preps | |
| FastRNA-24 Kits |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.