missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Dulbecco's MEM, w/ 4.5g/L Glucose Without L-Glut
Also known as DME, Dulbecco's Modified Eagle's Medium is the most widely used modification of Eagle's Basal Medium (BME).
Supplier: MP Biomedicals™ 091233254
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
DMEM Media | |
Glucose | |
500 mL | |
2° to 8°C |
Sterile | |
No Glutamine | |
6.9 to 7.4 |