missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ D-(+)- RAFFINOSE (POWDER)

Catalog No. mp114010032
Change view
Click to view available options
Quantity:
1000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114010032 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114010032 Supplier MP Biomedicals™ Supplier No. 114010032
Only null left

Highly pure, white crystalline powder with applications in cryopreservation

D-(+)- Raffinose is a trisaccharide composed of galactose, fructose and glucose which can be hydrolyzed to D-galactose and sucrose by the enzyme α-galactosidase (α-GAL), an enzyme not found in the human digestive tract. α-GAL also hydrolyzes other α-galactosides such as stachyose, verbascose, andgalactinol, if present. The enzyme does not cleave β-linked galactose, as in lactose.The raffinose family of oligosaccharides (RFOs) are alpha-galactosyl derivatives of sucrose, and the most common are the trisaccharideraffinose, the tetrasaccharide stachyose and the pentasaccharide verbascose.

  • Purity:≥98%
  • Physical appearance: White Crystalline powder
  • Synonyms: O-α-D-Galactopyranosyl-(1--->6)-α-D-glucopyranosyl β-D-fructofuranoside; Melitose; Melitriose
  • CAS Number: 17629-30-0
  • Molecular Formula: C18H32O16 5H2O
  • Molecular Weight: 594.5
  • Beilstein Registry Number: 99541
  • MDL Number: MFCD00071590
  • Optical Rotation: 101-107°

  • Used in procedures concerning cryopreservation to provide hypertonicity for cell desiccation prior to freezing)
  • Used as base substance for sucralose

Specifications

Content And Storage 15° to 30°C
Quantity 1000 g