missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Chymostatin, Microbial
Specifications
Specifications
| Concentration | 10mg/mL |
| Molecular Weight (g/mol) | 605.04 |
| Name | Chymostatin from Microbial |
| Quantity | 5 mg |
| Source | Microbial |
| Storage Requirements | 0°C |
| Form | Powder |