missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Bovine Fibrinogen

Suitable as a substrate for cell culture and in clotting assays

Supplier:  MP Biomedicals™ 08820224

 View more versions of this product

Catalog No. ICN820224

Only null left
Explore available promotions

Description

Description

Prepared by extensive washing of cryoprecipitate followed by a buffer extraction to remove unwanted proteins and precipitation with ammonium sulfate. Lyophilized powder. Clottable protein >90%.

Physical Characteristics

  • Appearance: Off-white powder
  • Clottable Protein (anhydrous): ≥90%
  • Moisture: ≤5%
  • Reported: Total protein, pH
Certifications: ISO, cGMP

Specifications

Specifications

Mammalian Cell Culture
6.4
Bovine Plasma
Lyophilized
Bovine Fibrinogen 90% Clottable from Bovine Plasma
5g
<-10°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions