missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Avidin from Egg White
Description
Avidin was used in an assay using functionalized xenon as a biosensor to detect biotin-avidin binding.
- CAS Number: 1405-69-2
- EC Number: 215-783-6
- MDL Number: MFCD00130572
- Format: White Crystalline Powder
- Specific Activity: Approx. 10 to 15 units/mg protein
- Soluble in water or dilute aqueous buffer
- Isoelectric point: 10
Specifications
Specifications
| Quantity | 100 mg |
| Source | Chicken |
| Content And Storage | 0°C |
| Form | Lyophilized |
| For Use With (Application) | Molecular Biology |
| Product Type | Avidin from Egg White |