missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ (Arg8)-Vasopressin
Supplier: MP Biomedicals™ 0215296205
Description
- Contains a disulfide bond between Cys1 and Cys6
Specifications
| Cell Biology and Analysis | |
| (Arg-8)-Vasopressin (AVP) | |
| -20°C |
| 1084.23 | |
| 5 mg |