missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Aprotinin, From Bovine Lung
Description
Aprotinin, [9087-70-1]
- C284H432N84O79S7
- Lyophilized white powder
Specifications
Specifications
| For Use With (Application) | Inhibition Assay |
| Molecular Weight (g/mol) | 6511.23 |
| Name | Aprotinin from Bovine Lung |
| pH Range | 5 to 7 |
| Quantity | 5 mg |
| Source | Bovine Lung |
| Storage Requirements | 4°C |
| Form | Lyophilized |