missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Aprotinin, From Bovine Lung

Catalog No. ICN19115801
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
100 mg
250 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19115801 1 mg
ICN19115805 5 mg
ICN19115810 10 mg
ICN19115825 25 mg
ICN19115880 100 mg
ICN19115883 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN19115801 Supplier MP Biomedicals™ Supplier No. 0219115801
Only null left

From Bovine Lung

Aprotinin, [9087-70-1]

  • C284H432N84O79S7
  • Lyophilized white powder

Specifications

For Use With (Application) Inhibition Assay
Molecular Weight (g/mol) 6511.23
Name Aprotinin from Bovine Lung
pH Range 5 to 7
Quantity 1 mg
Source Bovine Lung
Storage Requirements 4°C
Form Lyophilized