missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Aprotinin, Cell Culture Reagent
From bovine lung
Supplier: MP Biomedicals™ 0219455983
Description
Activity: ∽4 Inhibitor units/mg of dry weight. One Inhibitor unit = 1500 KIU. Lyophilized white powder.
Specifications
| Molecular Biology, Radioimmunoassays | |
| Aprotinin from Bovine Lung | |
| 250 mg | |
| 4°C |
| 6511.23 | |
| 5 to 7 | |
| Bovine Lung | |
| Lyophilized |