missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Apoferritin from Horse Spleen

Catalog No. ICN10026080
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10026080 100 mg
ICN10026025 25 mg
ICN10026090 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10026080 Supplier MP Biomedicals™ Supplier No. 0210026080
Only null left

Protein shell of ferritin, abundant in beta-cells of pancreas

When added to cultured endothelial cells apoferritin is taken up in a dose-responsive manner and appears to protect the cells from oxidant-mediated cytolysis.

  • CAS Number: 9013-31-4
  • MDL Number: MFCD00081365
  • Format: Off-white lyophilized powder
  • Protein Content: ≈90%
  • Iron Content: Typically <0.01%
  • Soluble in water or dilute buffer

Used for calibration of gel filtration columns and sodium dodecyl sulfate (SDS)-polyacrylamide gel electrophoresis. It acts as an anti-oxidant.

Specifications

For Use With (Application) Molecular Biology
Molecular Weight (g/mol) 445000
Name Apoferritin from Horse Spleen
Quantity 100 mg
Source Horse Spleen
Storage Requirements -20°C
Form Lyophilized