missing translation for 'onlineSavingsMsg'
Learn More

Moxifloxacin Hydrochloride Monohydrate 98.0+%, TCI America™

$190.13 - $1334.67

Chemical Identifiers

CAS 192927-63-2
Molecular Formula C21H27ClFN3O5
Molecular Weight (g/mol) 455.91
MDL Number MFCD09836237
InChI Key SKZIMSDWAIZNDD-UHFFFAOYNA-N
Synonym 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic Acid Hydrochloride
PubChem CID 9890250
IUPAC Name 1-cyclopropyl-6-fluoro-8-methoxy-7-{octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl}-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrate hydrochloride
SMILES O.Cl.COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CC2CCCNC2C1)C1CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M2479100MG
View Documents
TCI America
M2479100MG
100 mg
Each for $190.13
Only null left
 
M24791G
View Documents
TCI America
M24791G
1 g
Each for $1,334.67
Only null left
 
Description

Description

Specifications

Chemical Identifiers

192927-63-2
455.91
SKZIMSDWAIZNDD-UHFFFAOYNA-N
9890250
O.Cl.COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CC2CCCNC2C1)C1CC1
C21H27ClFN3O5
MFCD09836237
1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic Acid Hydrochloride
1-cyclopropyl-6-fluoro-8-methoxy-7-{octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl}-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrate hydrochloride

Specifications

192927-63-2
C21H27ClFN3O5
100 mg
SKZIMSDWAIZNDD-UHFFFAOYNA-N
1-cyclopropyl-6-fluoro-8-methoxy-7-{octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl}-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrate hydrochloride
9890250
≥98.0% (HPLC,T)
Moxifloxacin Hydrochloride Monohydrate
240°C
MFCD09836237
1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aS,7aS)-octahydro-1H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic Acid Hydrochloride
O.Cl.COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CC2CCCNC2C1)C1CC1
455.91
437.90
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : VB1983750

TSCA : No

SDS
Documents

Documents