missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse VEGF-165 Recombinant Protein, PeproTech®
Recombinant Protein
$132.00 - $6320.00
Specifications
| Accession Number | Q00731 |
|---|---|
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 22339 |
| Molecular Weight (g/mol) | 39 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
45032100UG
|
Gibco™
LS45032100UG |
100 μg |
Each for $1,268.00
|
|
|||||
4503210UG
|
Gibco™
LS4503210UG |
10 μg |
Each for $317.00
|
|
|||||
450321MG
|
Gibco™
LS450321MG |
1 mg |
Each for $6,320.00
|
|
|||||
45032250UG
|
Gibco™
45032250UG |
250 μg |
Each for $2,118.20
|
|
|||||
450322UG
|
Gibco™
LS450322UG |
2 μg |
Each for $132.00
|
|
|||||
45032500UG
|
Gibco™
45032500UG |
500 μg |
Each for $3,617.60
|
|
|||||
4503250UG
|
Gibco™
4503250UG |
50 μg |
Each for $723.52
|
|
|||||
Description
Recombinant Murine VEGF is a 39.0 kDa disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The vascular endothelial growth factor (VEGF) family currently includes VEGF-A, VEGF-B, VEGF-C, VEGF-D, VEGF-E, and PIGF. VEGF and its receptor system have been shown to be the fundamental regulators in the cell signaling of angiogenesis. Most tumors have the absolute requirement of angiogenesis and VEGF has been described as the most potent angiogenic cytokine linked to this process. To date 5 different isoforms of VEGF have been described. These isoforms are generated as the result of alternative splicing from a single VEGF gene. These various isoforms have been shown to bind to two tyrosine-kinase receptors flt-1 (VEGFR-1) and flk-1/KDR (VEGFR-2), which have been found to be expressed almost exclusively on endothelial cells.Specifications
| Q00731 | |
| protein with no preservative | |
| 39 kDa | |
| E. coli | |
| <1 EU/ μg | |
| VEGF-165 | |
| Determined by the dose-dependent stimulation of the proliferation of human umbilical vein endothelial cells (HUVEC) using a concentration range of 1.0-5.0 ng/ml | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| ELISA, Functional Assay, Western Blot | |
| 22339 | |
| Mouse VEGF-165 | |
| RUO | |
| H-VEGF-165; MVCD1; M-VEGF-165; vascular endothelial growth factor 165 b; vascular endothelial growth factor A; vascular endothelial growth factor A121; vascular endothelial growth factor A165; vascular permeability factor; VEGF; VEGF164; VEGF165; Vegfa; VEGF-A; VPF | |
| VEGFA | |
| Unconjugated | |
| MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKHCEPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR | |
| E. coli | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |