missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse VEGF-165, Animal-Free Recombinant Protein, PeproTech®
Description
Recombinant Murine VEGF 165 is a 39.0 kDa, disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The vascular endothelial growth factor (VEGF) family currently includes VEGF-A, VEGF-B, VEGF-C, VEGF-D, VEGF-E, and PIGF. VEGF and its receptor system have been shown to be the fundamental regulators in the cell signaling of angiogenesis. Most tumors have the absolute requirement of angiogenesis and VEGF has been described as the most potent angiogenic cytokine linked to this process. To date 5 different isoforms of VEGF have been described. These isoforms are generated as the result of alternative splicing from a single VEGF gene. These various isoforms have been shown to bind to two tyrosine-kinase receptors flt-1 (VEGFR-1) and flk-1/KDR (VEGFR-2), which have been found to be expressed almost exclusively on endothelial cells.
Specifications
Specifications
| Accession Number | Q00731 |
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 22339 |
| Molecular Weight (g/mol) | 39 kDa |
| Name | Mouse VEGF-165, Animal-Free |
| Purification Method | HPLC, SDS-PAGE |
| Quantity | 250 μg |
| Endotoxin Concentration | <0.1 EU/ μg |
| Show More |