missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse TNF-alpha Recombinant Protein, PeproTech®
Recombinant Protein
$131.00 - $4870.00
Specifications
| Accession Number | P06804 |
|---|---|
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 21926 |
| Molecular Weight (g/mol) | 17.3 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
31501A100UG
|
Gibco™
LS31501A100UG |
100 μg |
Each for $976.00
|
|
|||||
31501A1MG
|
Gibco™
LS31501A1MG |
1 mg |
Each for $4,870.00
|
|
|||||
31501A20UG
|
Gibco™
LS31501A20UG |
20 μg |
Each for $317.00
|
|
|||||
31501A250UG
|
Gibco™
31501A250UG |
250 μg |
Each for $1,622.21
|
|
|||||
31501A500UG
|
Gibco™
31501A500UG |
500 μg |
Each for $2,779.84
|
|
|||||
31501A50UG
|
Gibco™
LS31501A50UG |
50 μg |
Each for $584.00
|
|
|||||
31501A5UG
|
Gibco™
LS31501A5UG |
5 μg |
Each for $131.00
|
|
|||||
Description
Recombinant Murine TNF-alpha is a soluble 156 amino acid protein (17.3 kDa) which corresponds to C-terminal extracellular domain of the full length transmembrane protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.Specifications
| P06804 | |
| protein with no preservative | |
| 17.3 kDa | |
| E. coli | |
| <1 EU/ μg | |
| TNF alpha | |
| The ED50 as determined by the cytolysis of murine L929 cells in the presence of actinomycin D is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 10^7 units/mg | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| ELISA, Functional Assay, Western Blot | |
| 21926 | |
| Mouse TNF-alpha | |
| RUO | |
| APC1 protein; Cachectin; C-domain 1; C-domain 2; cTNF; DADB-70P7.1; DIF; ICD1; ICD2; Intracellular domain 1; Intracellular domain 2; N-terminal fragment; NTF; RATTNF; Tnf; TNF alpha; TNF superfamily; TNF a; TNF, macrophage-derived; TNF, monocyte-derived; TNFA; TNF-a; TNFalpha; TNF-alpha; Tnfsf1a; TNFSF2; TNFa; TNLG1F; Tumor necrosis factor; tumor necrosis factor (TNF superfamily, member 2); tumor necrosis factor alpha; tumor necrosis factor alpha (cachetin); tumor necrosis factor alpha precursor; tumor necrosis factor ligand 1 F; tumor necrosis factor ligand superfamily member 2; Tumor necrosis factor, membrane form; Tumor necrosis factor, soluble form; tumor necrosis factor-alpha; tumor necrosis factor-alpha precursor; tumor-necrosis factor; tumour necrosis factor | |
| Tnf | |
| Unconjugated | |
| MLRSSSQNSS DKPVAHVVAN HQVEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLVY SQVLFKGQGC PDYVLLTHTV SRFAISYQEK VNLLSAVKSP CPKDTPEGAE LKPWYEPIYL GGVFQLEKGD QLSAEVNLPK YLDFAESGQV YFGVIAL | |
| E. coli | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |