missing translation for 'onlineSavingsMsg'
Learn More

Mouse Siglec-E Alexa Fluor 405-conjugated Antibody, R&D Systems™

Catalog No. FAB5806V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5806V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB5806V100 Supplier Bio-Techne Supplier No. FAB5806V100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

Siglec-E Monoclonal specifically detects Siglec-E in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Siglec-E
Applications Flow Cytometry
Classification Monoclonal
Clone 750620
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q91Y57
Gene Symbols Siglece
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Siglec-E Gln19-Phe354 Accession # Q91Y57
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8778
Test Specificity Detects mouse Siglec-E in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Siglec-3/CD33 or rmSiglec-F is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG2b
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)