Learn More
Invitrogen™ Mouse ShhN (SHH)-Cell Lysates ELISA Kit
Description
The Mouse ShhN (SHH) ELISA quantitates Ms ShhN in mouse cell or tissue lysates. The assay will exclusively recognize both natural and recombinant Ms ShhN. Principle of the method The Mouse ShhN solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | Q62226 |
| Assay Range | 4.92-1200 pg/mL |
| Assay Sensitivity | 5 pg/mL |
| Calibration Range | 5 pg/mL to 1200 pg/mL |
| Content And Storage | Pre-coated 96-well plate, standard, detector antibody, streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers. May be stored at 2–8°C for up to 6 months from the date of shipment. |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Protein Family | Other Proteins |
| Show More |