missing translation for 'onlineSavingsMsg'
Learn More

Mouse SF20/MYDGF Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Catalog No. IC1104T100U
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1104T100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC1104T100U Supplier Bio-Techne Supplier No. IC1104T100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Monoclonal Antibody

SF20/MYDGF Monoclonal specifically detects SF20/MYDGF in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen SF20/MYDGF
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 2562A
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9CPT4
Gene Alias C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL25, IL27, IL27w, interleukin 27 working designation, interleukin-25, Ly6elg, MYDGF, myeloid-derived growth factor, R33729_1, SF20, Stromal cell-derived growth factor SF20, UPF0556 protein C19orf10
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived mouse SF20/MYDGF, Val25-Leu166, Accession # Q9CPT4
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56005
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Product Type Antibody
Form Purified
Isotype IgG
Product Line R&D Systems
Show More Show Less