missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse SF-20 Recombinant Protein, PeproTech®

Catalog No. 210252UG
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
250 μg
500 μg
1 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
210252UG 2 μg
21025100UG 100 μg
2102510UG 10 μg
210251MG 1 mg
21025250UG 250 μg
21025500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 210252UG Supplier Gibco™ Supplier No. LS210252UG
Only null left

Recombinant Protein

Recombinant Murine SF-20 is a 15.8 kDa protein containing 143 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The interleukins are a broad family of well characterized cytokines, primarily of hematopoietic cell origin. Cytokines are small, soluble proteins with pleiotropic effects on a variety of cell types. Cytokines have a regulatory function over the immune system and mediate aspects of inflammatory response. IL-25 (also called SF20/IL-25) is a secreted bone marrow stroma-derived growth factor cytokine that is derived from Th2 T-cells. IL-25 belongs to the IL-17 cytokine family and is capable of amplifying allergic type inflammatory responses by its actions on other cell types. IL-25 binds to mouse thymic shared antigen-1 and supports lymphoid cell proliferation. Infusion of mice with IL-25 induces IL-4, IL-5, and IL-13 gene expression. IL-25 induced mice also develop epithelial cell hyperplasia, increased mucus secretion, and airway hyperreactivity, suggesting that IL-25 may be an important mediator of allergic disease via production of IL-4, IL-5, IL-13 and eotaxin.

Specifications

Accession Number Q9CPT4
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 28106
Molecular Weight (g/mol) 15.8 kDa
Name Mouse SF-20
Quantity 2 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C19orf10; CS010; D17Wsu104e; EUROIMAGE1875335; IL 25; IL 27; Il25; IL27; IL27w; interleukin 25; interleukin 27 working designation; Interleukin25; interleukin-25; LOC501282; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; R33729 1; R33729_1; R337291; SF20; similar to lymphocyte antigen 6 complex, locus E ligand; stromal cell-derived growth factor SF20; UPF0556 protein C19orf10; UPF0556 protein C19orf10 homolog
Common Name C19orf10
Gene Symbol MYDGF
Conjugate Unconjugated
Recombinant Recombinant
Sequence MVSEPTTVPF DVRPGGVVHS FSQDVGPGNK FTCTFTYASQ GGTNEQWQMS LGTSEDSQHF TCTIWRPQGK SYLYFTQFKA ELRGAEIEYA MAYSKAAFER ESDVPLKSEE FEVTKTAVSH RPGAFKAELS KLVIVAKAAR SEL
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less